Learn More
SPEX CERTIPREP LLC 3P SULFATE STD 100PPM/100ML
Catalog No. NC9799489
Encompass_Preferred
(HPS by Zeptometrix) 100 ug/mL Sulfate from Na2SO4 in water, 99.99+% (metals basis); Single Component IC Standard, 18 Month Expiry Date. Prepared from high-purity salts in 18-megaohm water, packaged in HDPE bottles.
Catalog No. NC9799489 Supplier SPEX CERTIPREP LLC 3P Supplier No. ICSS100
(HPS by Zeptometrix) 100 ug/mL Sulfate from Na2SO4 in water, 99.99+% (metals basis); Single Component IC Standard, 18 Month Expiry Date. Prepared from high-purity salts in 18-megaohm water, packaged in HDPE bottles.