Learn More
Hach Company Sulfate TNTplus Vial Test, HR (150-900 mg/L SO₄), 25 Tests
Catalog No. 501998150 Shop All Hach Company Products
Encompass
Sulfate TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Sulfate by the Turbidimetric Method.
Catalog No. 50-199-8150 Supplier Hach Company Supplier No. TNT865
Sulfate TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Sulfate by the Turbidimetric Method.