Learn More
Medchemexpress LLC Sulfathiazole | 72-14-0 | MFCD00005319 | 99.7% | 1 G
SDP
Catalog No. 5000409488
Encompass_Preferred
Sulfathiazole is an orally active endocrine disruptor that targets the steroidogenic pathway. It specifically enhances the activity of CYP19 in human adrenal cancer cells (H295R) and upregulates the mRNA expression of CYP17, CYP19, and 3β-HSD. It also increases the production of 17-estradiol (E2) and demonstrates endocrine disrupting effects on aquatic organisms like the Japanese medaka fish.
  • Targets the steroidogenic pathway
  • Enhances CYP19 activity in human adrenal cancer cells (H295R)
  • Upregulates mRNA expression of CYP17, CYP19, and 3β-HSD
  • Increases production of 17-estradiol (E2)
  • Demonstrates endocrine disrupting effects on aquatic organisms
  • Can inhibit copper corrosion in 0.1 M NaCl solution
  • Increases plasma 17-estradiol concentrations in male Japanese medaka models
Catalog No. 50-004-09488 Supplier Medchemexpress LLC Supplier No. HYB05071G
Sulfathiazole is an orally active endocrine disruptor that targets the steroidogenic pathway. It specifically enhances the activity of CYP19 in human adrenal cancer cells (H295R) and upregulates the mRNA expression of CYP17, CYP19, and 3β-HSD. It also increases the production of 17-estradiol (E2) and demonstrates endocrine disrupting effects on aquatic organisms like the Japanese medaka fish.
  • Targets the steroidogenic pathway
  • Enhances CYP19 activity in human adrenal cancer cells (H295R)
  • Upregulates mRNA expression of CYP17, CYP19, and 3β-HSD
  • Increases production of 17-estradiol (E2)
  • Demonstrates endocrine disrupting effects on aquatic organisms
  • Can inhibit copper corrosion in 0.1 M NaCl solution
  • Increases plasma 17-estradiol concentrations in male Japanese medaka models