Learn More
Medchemexpress LLC Sulfathiazole-d4 | 1020719-89-4 | 99.9% | 5 MG
SDP
Catalog No. 5000408035
Encompass_Preferred
Sulfathiazole-d4 is a deuterium labeled Sulfathiazole. Sulfathiazole is an orally active, endocrine disruptor targeting the steroidogenic pathway, specifically enhancing the activity of CYP19 in human adrenal cancer cells (H295R) and upregulating the mRNA expression of CYP17, CYP19, and 3β-HSD. It increases the production of 17-estradiol (E2) and has endocrine disrupting effects on aquatic organisms such as the Japanese medaka fish.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Enhances the activity of CYP19 in human adrenal cancer cells (H295R)
  • Upregulates the mRNA expression of CYP17, CYP19, and 3β-HSD
  • Increases the production of 17-estradiol (E2)
  • Has endocrine disrupting effects on aquatic organisms such as the Japanese medaka fish
Catalog No. 50-004-08035 Supplier Medchemexpress LLC Supplier No. HYB0507S5MG
Sulfathiazole-d4 is a deuterium labeled Sulfathiazole. Sulfathiazole is an orally active, endocrine disruptor targeting the steroidogenic pathway, specifically enhancing the activity of CYP19 in human adrenal cancer cells (H295R) and upregulating the mRNA expression of CYP17, CYP19, and 3β-HSD. It increases the production of 17-estradiol (E2) and has endocrine disrupting effects on aquatic organisms such as the Japanese medaka fish.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Enhances the activity of CYP19 in human adrenal cancer cells (H295R)
  • Upregulates the mRNA expression of CYP17, CYP19, and 3β-HSD
  • Increases the production of 17-estradiol (E2)
  • Has endocrine disrupting effects on aquatic organisms such as the Japanese medaka fish