Learn More
Apexbio Technology LLC SULFATHIAZOLE-SODIUM-10G
SDP
Catalog No. 5000568147
Encompass_Preferred
Sulfathiazole sodium (CAS No 144-74-1) is commonly used in biomedical research as an antibacterial agent belonging to the sulfonamide class Originating from synthetic sulfonamide compounds it works by inhibiting bacterial folic acid synthesis through competitive antagonism of para-aminobenzoic acid (PABA) in the dihydropteroate synthase enzyme pathway Its in vitro activity is generally observed in the micromolar range against gram-positive and gram-negative bacteria It is frequently applied in various experimental models including cell cultures and animal studies to evaluate antibacterial efficacy and to screen for potential therapeutic effects Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-68147 Supplier Apexbio Technology LLC Supplier No. B343210000
Sulfathiazole sodium (CAS No 144-74-1) is commonly used in biomedical research as an antibacterial agent belonging to the sulfonamide class Originating from synthetic sulfonamide compounds it works by inhibiting bacterial folic acid synthesis through competitive antagonism of para-aminobenzoic acid (PABA) in the dihydropteroate synthase enzyme pathway Its in vitro activity is generally observed in the micromolar range against gram-positive and gram-negative bacteria It is frequently applied in various experimental models including cell cultures and animal studies to evaluate antibacterial efficacy and to screen for potential therapeutic effects Experimental concentrations typically depend on the specific research design