Learn More
Apexbio Technology LLC SULFATHIAZOLE-SODIUM-5G
SDP
Catalog No. 5000569081
Encompass_Preferred
Sulfathiazole sodium (CAS No 144-74-1) is commonly used in biomedical research as an antibacterial agent belonging to the sulfonamide class Originating from synthetic sulfonamide compounds it works by inhibiting bacterial folic acid synthesis through competitive antagonism of para-aminobenzoic acid (PABA) in the dihydropteroate synthase enzyme pathway Its in vitro activity is generally observed in the micromolar range against gram-positive and gram-negative bacteria It is frequently applied in various experimental models including cell cultures and animal studies to evaluate antibacterial efficacy and to screen for potential therapeutic effects Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-69081 Supplier Apexbio Technology LLC Supplier No. B34325000
Sulfathiazole sodium (CAS No 144-74-1) is commonly used in biomedical research as an antibacterial agent belonging to the sulfonamide class Originating from synthetic sulfonamide compounds it works by inhibiting bacterial folic acid synthesis through competitive antagonism of para-aminobenzoic acid (PABA) in the dihydropteroate synthase enzyme pathway Its in vitro activity is generally observed in the micromolar range against gram-positive and gram-negative bacteria It is frequently applied in various experimental models including cell cultures and animal studies to evaluate antibacterial efficacy and to screen for potential therapeutic effects Experimental concentrations typically depend on the specific research design