Learn More
Cayman Chemical SulfatIdebovInsodIum sa 10mg
SDP
Catalog No. 502935180
Encompass_Preferred
A mixture of isolated bovine sulfatides; found in the central and peripheral nervous systems, pancreas, and immune system; primarily found in myelin; the ceramide portion contains variable fatty acid chain lengths, which are tissue- and pathology-dependent; accumulate in the lysosome of patients with metachromatic leukodystrophy
Catalog No. 50-293-5180 Supplier Cayman Chemical Supplier No. 2432310 MG
A mixture of isolated bovine sulfatides; found in the central and peripheral nervous systems, pancreas, and immune system; primarily found in myelin; the ceramide portion contains variable fatty acid chain lengths, which are tissue- and pathology-dependent; accumulate in the lysosome of patients with metachromatic leukodystrophy