Learn More
Cayman Chemical Sulfatide (bovine) (sodium salt), Purity: ≥95%, 50 mg
SDP
Catalog No. 502935181
Encompass_Preferred
A mixture of isolated bovine sulfatides; found in the central and peripheral nervous systems, pancreas, and immune system; primarily found in myelin; the ceramide portion contains variable fatty acid chain lengths, which are tissue- and pathology-dependent; accumulate in the lysosomes of patients with metachromatic leukodystrophy; located in pancreatic β-cells and inhibit insulin release from isolated rat pancreatic islet cells; brain levels are decreased in patients with preclinical Alzheimer's disease; autoantibodies against sulfatide have been found in CSF of patients with multiple sclerosis; intratumoral levels are positively correlated with increased tumor stage in patients with ovarian cancer
Catalog No. 50-293-5181 Supplier Cayman Chemical Supplier No. 2432350 MG
A mixture of isolated bovine sulfatides; found in the central and peripheral nervous systems, pancreas, and immune system; primarily found in myelin; the ceramide portion contains variable fatty acid chain lengths, which are tissue- and pathology-dependent; accumulate in the lysosomes of patients with metachromatic leukodystrophy; located in pancreatic β-cells and inhibit insulin release from isolated rat pancreatic islet cells; brain levels are decreased in patients with preclinical Alzheimer's disease; autoantibodies against sulfatide have been found in CSF of patients with multiple sclerosis; intratumoral levels are positively correlated with increased tumor stage in patients with ovarian cancer