Learn More
Sigma Aldrich Fine Chemicals Biosciences Sulfatides from bovine bra5MG
Catalog No. 502906487
Encompass
Sulfatide is a 3-O-sulfogalactosylceramide consisting primarily of galacto cerebroside sulfate. It is localized in myelin sheath and comprises approximately 4% of total myelin lipid in brain.
Catalog No. 50-290-6487 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S10065MG
Sulfatide is a 3-O-sulfogalactosylceramide consisting primarily of galacto cerebroside sulfate. It is localized in myelin sheath and comprises approximately 4% of total myelin lipid in brain.