Learn More
Selleck Chemical LLC Sulfatinib S0487-5MG
SDP
Supplier:  Selleck Chemical LLC S04875MG
Encompass_Preferred
Sulfatinib is a potent and highly selective tyrosine kinase inhibitor against VEGFR1 VEGFR2 VEGFR3 FGFR1 and CSF1R with IC50 of 2 nM 24 nM 1 nM 15 nM and 4 nM respectively Sulfatinib shows encouraging antitumor activity and manageable toxicities in patients with advanced NETs
Catalog No. 50-217-3040