Learn More
Hach Company SulfaVer 4 Sulfate Reagent Powder Pillows, 25-mL, pk/100
Catalog No. 501994851 Shop All Hach Company Products
Encompass
For sulfate determination by the SulfaVer 4 Turbidimetric Method. USEPA accepted for reporting wastewater analysis using Hach Method 8051. Range: up to 70 mg/L. Sample Size: 25 mL. Pack of 100 powder pillows. To confirm accurate results, see appropriate standard solutions on the Consumables tab.
Catalog No. 50-199-4851 Supplier Hach Company Supplier No. 1206599
For sulfate determination by the SulfaVer 4 Turbidimetric Method. USEPA accepted for reporting wastewater analysis using Hach Method 8051. Range: up to 70 mg/L. Sample Size: 25 mL. Pack of 100 powder pillows. To confirm accurate results, see appropriate standard solutions on the Consumables tab.