Learn More
Hach Company SulfaVer 4 Sulfate Reagent Powder Pillows, 10 mL, pk/100
Catalog No. 501995411 Shop All Hach Company Products
Encompass
For determination of sulfate by the SulfaVer 4 turbidimetric method. USEPA accepted for reporting wastewater analysis using Hach Method 8051. Method: SulfaVer 4. Range: 2 to 70 mg/L SO4. Sample Size: 10 mL. Pack of 100 Powder Pillows. To confirm accurate results, see appropriate standard solutions on the Consumables tab.
Catalog No. 50-199-5411 Supplier Hach Company Supplier No. 2106769
For determination of sulfate by the SulfaVer 4 turbidimetric method. USEPA accepted for reporting wastewater analysis using Hach Method 8051. Method: SulfaVer 4. Range: 2 to 70 mg/L SO4. Sample Size: 10 mL. Pack of 100 Powder Pillows. To confirm accurate results, see appropriate standard solutions on the Consumables tab.