Learn More
Hach Company Sulfide 1 Reagent, 100 mL MDB
Supplier:  Hach Company 181632
Encompass
For sulfide determination by the Methylene Blue method (Hach method 8131). Range: up to 800 μg/L. Also used with visual test kits. 100 mL Marked Dropping Bottle
Catalog No. 50-199-4608