Learn More
Hach Company Sulfide 1 Reagent, 500 mL
Supplier:  Hach Company 181649
Encompass
For sulfide determination by the Methylene Blue method (Hach method 8131). Range: up to 800 μg/L. Also used with visual test kits. 500 mL bottle
Catalog No. 50-199-4609