Learn More
Hach Company Sulfide 2 Reagent, 100 mL MDB
Supplier: Hach Company 181732
For sulfide determination by the Methylene Blue method (Hach method 8131). Range: up to 800 μg/L. Also used with visual test kits. 100 mL Marked Dropping Bottle