Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics Sulfide Activator Solution
SDP

Catalog No. 13299217J
Change view
Click to view available options
Quantity:
6 x 10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-299-217J 6 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-299-217J Supplier CHEMetrics Supplier No. A9500
Add to Cart
Add to Cart

The Sulfide Activator Solution accessory pack (A-9500) supplies enough solution to perform at least 200 tests.

Specifications

Format Liquid
For Use With (Equipment) Sulfide CHEMets, Vacu-vials and VACUettes test kit
No. per Pack 6
Quantity 6 x 10 mL
Type Sulfide Activator Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.