Learn More
Aqua Solutions Sulfide Standard (APHA) 1,000 ppm w/w (1 ml = 1 mg S-²) (500ML)
SDP
Catalog No. NC9713472
Encompass_Preferred
Sulfide Standard (APHA) 1,000 ppm w/w (1 ml = 1 mg S-²) (500ML)
*Solution expires at 2 months. from date of manufacture
Catalog No. NC9713472 Supplier Aqua Solutions Supplier No. 8975500ML
Sulfide Standard (APHA) 1,000 ppm w/w (1 ml = 1 mg S-²) (500ML)
*Solution expires at 2 months. from date of manufacture