Learn More
Aqua Solutions Sulfide Standard (APHA) 1,000 ppm w/w (1 ml = 1 mg S-²) (100ML)
SDP
Catalog No. NC9639129
Encompass_Preferred
Sulfide Standard (APHA) 1,000 ppm w/w (1 ml = 1 mg S-²) (100ML)
*Solution expires at 2 months from date of manufacture.
Catalog No. NC9639129 Supplier Aqua Solutions Supplier No. 8975100ML
Sulfide Standard (APHA) 1,000 ppm w/w (1 ml = 1 mg S-²) (100ML)
*Solution expires at 2 months from date of manufacture.