Learn More
CHEMetrics VACUettes Sulfide Refill

Specifications
Specifications
| Analytes | Sulfide |
| Format | Liquid |
| For Use With (Equipment) | C-9501C and C-9510C comparators |
| Includes | 30 self-filling VACUettes ampoules to refill kit K-9510C |
| Quantity | 30 ampoules |
| Type | Sulfide VACUettes Refill |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.