Learn More
Medchemexpress LLC Sulfinpyrazone (Standard) | 57-96-5 | 96.8% | 25 MG
SDP
Catalog No. 5000414416
Encompass_Preferred
Sulfinpyrazone (Standard) is the analytical standard of Sulfinpyrazone, intended for research and analytical applications. It is an orally active and potent uricosuric agent for chronic and intermittent gouty arthritis, also possessing antithrombotic and platelet inhibitory effects. As an analytical standard, it is commonly used in qualitative, quantitative, and methodological research experiments.
  • Functions as an analytical standard
  • Suitable for research and analytical applications
  • Acts as an orally active and potent uricosuric agent
  • Used for chronic and intermittent gouty arthritis
  • Possesses antithrombotic and platelet inhibitory effects
  • Utilized in experiments like HPLC, GC, and MS
Catalog No. 50-004-14416 Supplier Medchemexpress LLC Supplier No. 25MGHYB1271R
Sulfinpyrazone (Standard) is the analytical standard of Sulfinpyrazone, intended for research and analytical applications. It is an orally active and potent uricosuric agent for chronic and intermittent gouty arthritis, also possessing antithrombotic and platelet inhibitory effects. As an analytical standard, it is commonly used in qualitative, quantitative, and methodological research experiments.
  • Functions as an analytical standard
  • Suitable for research and analytical applications
  • Acts as an orally active and potent uricosuric agent
  • Used for chronic and intermittent gouty arthritis
  • Possesses antithrombotic and platelet inhibitory effects
  • Utilized in experiments like HPLC, GC, and MS