Learn More
Medchemexpress LLC Sulfinpyrazone (Standard) | 57-96-5 | 96.8% | 250 MG
SDP
Catalog No. 5000414225
Encompass_Preferred
Sulfinpyrazone (Standard) is the analytical standard of Sulfinpyrazone. This product is intended for research and analytical applications. Sulfinpyrazone is an orally active and potent uricosuric agent for chronic and intermittent gouty arthritis, and has antithrombotic and platelet inhibitory effects.
  • Analytical standard grade
  • Reference standard supplied assay
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
Catalog No. 50-004-14225 Supplier Medchemexpress LLC Supplier No. 250MGHYB1271R
Sulfinpyrazone (Standard) is the analytical standard of Sulfinpyrazone. This product is intended for research and analytical applications. Sulfinpyrazone is an orally active and potent uricosuric agent for chronic and intermittent gouty arthritis, and has antithrombotic and platelet inhibitory effects.
  • Analytical standard grade
  • Reference standard supplied assay
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS