Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfisomidine, TCI America™
SDP

Catalog No. S03621G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03621G Supplier TCI America Supplier No. S03621G

Chemical Identifiers

CAS 515-64-0
Molecular Formula C12H14N4O2S
Molecular Weight (g/mol) 278.33
MDL Number MFCD00010567
InChI Key YZMCKZRAOLZXAZ-UHFFFAOYSA-N
PubChem CID 5343
ChEBI CHEBI:32166
IUPAC Name 4-amino-N-(2,6-dimethylpyrimidin-4-yl)benzenesulfonamide
SMILES CC1=CC(=NC(=N1)C)NS(=O)(=O)C2=CC=C(C=C2)N

Specifications

Type Sulfisomidine
Physical Form Crystalline Powder
Melting Point 245°C
Color White/Yellow
Quantity 1 g
Formula Weight 278.33
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material Sulfisomidine
RTECSNumber WO9270000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.