Learn More
Cayman Chemical Sulfisoxazole, Purity: ≥98%, CAS: 127-69-5, 10 g
SDP
Catalog No. 502935791
Encompass_Preferred
A sulfonamide antibiotic and nonpeptide ET receptor antagonist (IC50s = 0.6 and 22 μM for the ETA and ETB receptors, respectively); inhibits the growth of Gram-positive and Gram-negative bacteria in vitro, including S. pneumoniae, B. subtilis, S. epidermidis, E. coli, and K. pneumoniae (MICs = 1.95, 0.98, 7.81, 62.5, and 7.81 μg/mL, respectively); inhibits phosphoinositide turnover stimulated by ET-1 in TE671 cells; inhibits ET-induced contraction of rat pulmonary artery rings ex vivo at 0.1 and 1 mM; reduces atrial natriuretic peptide levels in myocytes and plasma and increases survival in a rat model of pulmonary hypertension induced by MCT at 1,000 mg/kg per day; blocks MCT-induced increases in pulmonary artery blood pressure at 300 and 1,000 mg/kg
Catalog No. 50-293-5791 Supplier Cayman Chemical Supplier No. 2598010 G
A sulfonamide antibiotic and nonpeptide ET receptor antagonist (IC50s = 0.6 and 22 μM for the ETA and ETB receptors, respectively); inhibits the growth of Gram-positive and Gram-negative bacteria in vitro, including S. pneumoniae, B. subtilis, S. epidermidis, E. coli, and K. pneumoniae (MICs = 1.95, 0.98, 7.81, 62.5, and 7.81 μg/mL, respectively); inhibits phosphoinositide turnover stimulated by ET-1 in TE671 cells; inhibits ET-induced contraction of rat pulmonary artery rings ex vivo at 0.1 and 1 mM; reduces atrial natriuretic peptide levels in myocytes and plasma and increases survival in a rat model of pulmonary hypertension induced by MCT at 1,000 mg/kg per day; blocks MCT-induced increases in pulmonary artery blood pressure at 300 and 1,000 mg/kg