Learn More
Medchemexpress LLC Sulfisoxazole | 127-69-5 | >99.9% | 250 MG
SDP
Catalog No. 5000406825
Encompass_Preferred
Sulfisoxazole (Sulfafurazole) is an orally active endothelin receptor antagonist with IC50 values of 0.60 μM and 22 μM against endothelin receptor A and endothelin receptor B, respectively. It is also a sulfonamide antibacterial agent with an oxazole substituent, known to inhibit breast cancer exosome release by targeting endothelin receptor A.
  • Orally active endothelin receptor antagonist
  • Inhibits endothelin receptor A and B
  • Sulfonamide antibacterial agent
  • Inhibits breast cancer exosome release
Catalog No. 50-004-06825 Supplier Medchemexpress LLC Supplier No. 250MGHYB0323R
Sulfisoxazole (Sulfafurazole) is an orally active endothelin receptor antagonist with IC50 values of 0.60 μM and 22 μM against endothelin receptor A and endothelin receptor B, respectively. It is also a sulfonamide antibacterial agent with an oxazole substituent, known to inhibit breast cancer exosome release by targeting endothelin receptor A.
  • Orally active endothelin receptor antagonist
  • Inhibits endothelin receptor A and B
  • Sulfonamide antibacterial agent
  • Inhibits breast cancer exosome release