Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfisoxazole, 99%, Thermo Scientific Chemicals

Catalog No. AC439670250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC439670250 25 g
AC439671000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC439670250 Supplier Thermo Scientific Chemicals Supplier No. 439670250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-69-5
Molecular Formula C11H13N3O3S
Molecular Weight (g/mol) 267.3
InChI Key NHUHCSRWZMLRLA-UHFFFAOYSA-N
Synonym sulfisoxazole, sulfafurazole, sulphafurazole, sulfisoxazol, sulfafurazol, sulfaisoxazole, sulfofurazole, sulfisoxasole, sulphaisoxazole, sulfadimethylisoxazole
PubChem CID 5344
ChEBI CHEBI:102484
IUPAC Name 4-amino-N-(3,4-dimethyl-1,2-oxazol-5-yl)benzenesulfonamide
SMILES CC1=C(ON=C1C)NS(=O)(=O)C2=CC=C(C=C2)N

Specifications

Melting Point 194°C
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Merck Index 15,9082
Solubility Information Solubility in water: 0.13mg/mL (25°C). Other solubilities: soluble in alcohol
Formula Weight 267.3
Percent Purity 99%
Chemical Name or Material Sulfisoxazole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 204-858-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.