Learn More
Hach Company Sulfite 1 Reagent Powder Pillows, pk/100
Supplier:  Hach Company 220399
Encompass
For determination of sulfite, chlorine and hydrogen peroxide by titration methods. Pack of 100 powder pillows.
Catalog No. 50-199-4718