Learn More
Hach Company Sulfite Standard Solution, 15 mg/L as SO₃ (equivalent), 500 mL
Catalog No. 501995926 Shop All Hach Company Products
Encompass
Sulfite equivalent standard solution. This equivalent standard is prepared with stabilized Sodium Thiosulfate, since sulfite is not a stable in solution. Equivalent to 15 mg/L as SO3. 500 mL Bottle
Catalog No. 50-199-5926 Supplier Hach Company Supplier No. 2408449
Sulfite equivalent standard solution. This equivalent standard is prepared with stabilized Sodium Thiosulfate, since sulfite is not a stable in solution. Equivalent to 15 mg/L as SO3. 500 mL Bottle