Learn More
Cederlane Sulfite, UV Method Test Kit
SDP
Supplier:  Cederlane AK00071
Encompass
Enzymatic method for the determination of sulfite. Based on the spectrophotometric measurement of NADH formed through the combined action of sulfite oxidase (SOD) and NADH-peroxidase (NADH-POD).
Catalog No. NC0260617