Learn More
AdipoGen Sulfluramid
SDP
Supplier:  AdipoGen CDXS0050M100
Encompass_Preferred
Chemical. CAS 4151-50-2. Formula C10H6F17NO2S. MW 527.2. Synthetic. Perfluorinated delayed-action insecticide/pesticide. Mode of action is by uncoupling oxidative phosphorylation via disruption of the proton gradient. Suppresses immunoglobulin Ig M production in mice. Shown to be cytotoxic. Compound can be used as analytical reference material.
Catalog No. 50-256-0079