Learn More
Abcam Sulfluramid, Perfluorinated pesticide, 250MG
Catalog No. 502706202
Encompass_Preferred
MW 527.2 Da, Purity >98%. Perfluorinated pesticide and delayed-action insecticide. Suppresses immunoglobulin (Ig) M production in mice. Cytotoxic effect by uncoupling oxidative phosphorylation. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6202 Supplier Abcam Supplier No. AB142699250MG
MW 527.2 Da, Purity >98%. Perfluorinated pesticide and delayed-action insecticide. Suppresses immunoglobulin (Ig) M production in mice. Cytotoxic effect by uncoupling oxidative phosphorylation. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement