Learn More
Medchemexpress LLC Sulfo-ara-F-NMN | 1374663-29-2 | 99.36% | 352.28 | 10 MG
SDP
Catalog No. 5000365600
Encompass_Preferred
Sulfo-ara-F-NMN (CZ-48) is a mimetic of nicotinamide mononucleotide (NMN). It acts selectively by activating SARM1 and inhibiting CD38 (IC50 around 10 μM), leading to the induction of intracellular cyclic ADP-ribose (cADPR) production.
  • Mimics nicotinamide mononucleotide
  • Selectively activates SARM1
  • Inhibits CD38 (IC50 around 10 μM)
  • Induces intracellular cyclic ADP-ribose production
  • For research use only
  • Store at -20°C under nitrogen
  • Unstable in solutions; use freshly prepared solutions
Catalog No. 50-003-65600 Supplier Medchemexpress LLC Supplier No. 10MGHY129522
Sulfo-ara-F-NMN (CZ-48) is a mimetic of nicotinamide mononucleotide (NMN). It acts selectively by activating SARM1 and inhibiting CD38 (IC50 around 10 μM), leading to the induction of intracellular cyclic ADP-ribose (cADPR) production.
  • Mimics nicotinamide mononucleotide
  • Selectively activates SARM1
  • Inhibits CD38 (IC50 around 10 μM)
  • Induces intracellular cyclic ADP-ribose production
  • For research use only
  • Store at -20°C under nitrogen
  • Unstable in solutions; use freshly prepared solutions