Learn More
Medchemexpress LLC Sulfo-ara-F-NMN | 1374663-29-2 | 99.4% | 25 MG
SDP
Catalog No. 5000339187
Encompass_Preferred
Sulfo-ara-F-NMN (CZ-48) is a mimetic of nicotinamide mononucleotide (NMN) that selectively activates SARM1 and inhibits CD38 with an IC50 of approximately 10 μM. This compound stimulates intracellular cyclic ADP-ribose (cADPR) production and subsequently activates SARM1 to generate cyclic ADP-ribose, leading to non-apoptotic cell death. It is intended for research use only.
  • Mimetic of nicotinamide mononucleotide (NMN)
  • Selectively activates SARM1
  • Inhibits CD38 (IC50 around 10 μM)
  • Induces intracellular cyclic ADP-ribose (cADPR) production
  • Activates SARM1 to produce cyclic ADP-ribose
  • Induces non-apoptotic cell death
  • For research use only
Catalog No. 50-003-39187 Supplier Medchemexpress LLC Supplier No. 25MGHY129522
Sulfo-ara-F-NMN (CZ-48) is a mimetic of nicotinamide mononucleotide (NMN) that selectively activates SARM1 and inhibits CD38 with an IC50 of approximately 10 μM. This compound stimulates intracellular cyclic ADP-ribose (cADPR) production and subsequently activates SARM1 to generate cyclic ADP-ribose, leading to non-apoptotic cell death. It is intended for research use only.
  • Mimetic of nicotinamide mononucleotide (NMN)
  • Selectively activates SARM1
  • Inhibits CD38 (IC50 around 10 μM)
  • Induces intracellular cyclic ADP-ribose (cADPR) production
  • Activates SARM1 to produce cyclic ADP-ribose
  • Induces non-apoptotic cell death
  • For research use only