Learn More
Vector Laboratories Sulfo ChromaLINK Biotin (Water Soluble), 5 x 1 mg
SDP
Catalog No. 502045720
Encompass
Bioconjugation. ChromaLINK Biotin contains a UV-traceable chromophore based on ChromaLINK technology to enable reproducibility in your biotinylation process. Now you can measure the degree of biotinylation in minutes, not hours, without the standard curves required for HABA/avidin and fluoro-reporter assays. With a simple, non-destructive UV scan, you can quantify biotin incorporation and ensure reproducible production of consistent batches. This biotinylation reagent is water soluble, which means no DMF or other organic solvent is required to dissolve the compound prior to labeling.
Catalog No. 50-204-5720 Supplier Vector Laboratories Supplier No. B1007105
Bioconjugation. ChromaLINK Biotin contains a UV-traceable chromophore based on ChromaLINK technology to enable reproducibility in your biotinylation process. Now you can measure the degree of biotinylation in minutes, not hours, without the standard curves required for HABA/avidin and fluoro-reporter assays. With a simple, non-destructive UV scan, you can quantify biotin incorporation and ensure reproducible production of consistent batches. This biotinylation reagent is water soluble, which means no DMF or other organic solvent is required to dissolve the compound prior to labeling.