Learn More
Medchemexpress LLC Sulfo-Cy3 amine | 2183440-43-7 | 99.66% | 714.93 | 100 MG
SDP
Catalog No. 5000428473
Encompass_Preferred
Sulfo-Cy3 amine is a dye derivative of Cyanine 3 (Cy3) that bears an amine group. The sulfonate ion enhances the compound's water solubility, making it suitable for aqueous solutions. Cy3 is a fluorescent dye that typically exhibits a fluorescence spectrum in the green to orange wavelength range. The amine functionality allows Sulfo-Cy3 amine to react with carboxyl groups to form covalent bonds.
  • Binds to biological molecules such as proteins and antibodies.
  • Enables tracking of location and dynamic changes within biological samples.
  • Enhanced water solubility.
  • Suitable for aqueous solutions.
Catalog No. 50-004-28473 Supplier Medchemexpress LLC Supplier No. HYD1272100MG
Sulfo-Cy3 amine is a dye derivative of Cyanine 3 (Cy3) that bears an amine group. The sulfonate ion enhances the compound's water solubility, making it suitable for aqueous solutions. Cy3 is a fluorescent dye that typically exhibits a fluorescence spectrum in the green to orange wavelength range. The amine functionality allows Sulfo-Cy3 amine to react with carboxyl groups to form covalent bonds.
  • Binds to biological molecules such as proteins and antibodies.
  • Enables tracking of location and dynamic changes within biological samples.
  • Enhanced water solubility.
  • Suitable for aqueous solutions.