Learn More
Medchemexpress LLC Sulfo-Cy3 amine | 2183440-43-7 | 99.7% | 714.93 g/mol | C36H50N4O7S2 | 10 MG
SDP
Catalog No. 5000296690
Encompass_Preferred
Sulfo-Cy3 amine is a water-soluble Cyanine3 derivative containing a primary amine for covalent conjugation to biomolecules. The sulfonate groups increase aqueous solubility and the dye provides bright, photostable fluorescence for imaging and detection applications. Soluble in DMSO; may require ultrasonication and dry DMSO for best results.
  • Water-soluble sulfonated Cy3 derivative suitable for aqueous labeling.
  • Primary amine functional group for conjugation to electrophiles and carboxyl groups.
  • High purity for consistent labeling performance.
  • Soluble in DMSO (125 mg/mL); may require ultrasonication and newly opened DMSO.
  • Bright, photostable fluorescence for imaging and detection applications.
Catalog No. 50-002-96690 Supplier Medchemexpress LLC Supplier No. HYD127210MG
Sulfo-Cy3 amine is a water-soluble Cyanine3 derivative containing a primary amine for covalent conjugation to biomolecules. The sulfonate groups increase aqueous solubility and the dye provides bright, photostable fluorescence for imaging and detection applications. Soluble in DMSO; may require ultrasonication and dry DMSO for best results.
  • Water-soluble sulfonated Cy3 derivative suitable for aqueous labeling.
  • Primary amine functional group for conjugation to electrophiles and carboxyl groups.
  • High purity for consistent labeling performance.
  • Soluble in DMSO (125 mg/mL); may require ultrasonication and newly opened DMSO.
  • Bright, photostable fluorescence for imaging and detection applications.