Learn More
Medchemexpress LLC Sulfo-Cy3 Azide | 1801695-56-6 | C35H46N6O10S3 | 1 MG
SDP
Catalog No. 5000372073
Encompass_Preferred
Sulfo-Cy3 Azide is a water-soluble fluorogenic dye featuring an azide group, which facilitates Click Chemistry. It is suitable for labeling delicate molecules, including proteins. The azide group allows it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules, as well as ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • Water-soluble fluorogenic dye: allows for use in aqueous environments.
  • Azide group: enables Click Chemistry reactions.
  • Versatile labeling: can be used to label sensitive molecules such as proteins.
  • Compatible with CuAAc: reacts with alkyne groups in copper-catalyzed azide-alkyne cycloaddition.
  • Compatible with SPAAC: undergoes ring strain-promoted alkyne-azide cycloaddition with DBCO or BCN groups.
Catalog No. 50-003-72073 Supplier Medchemexpress LLC Supplier No. HY1517101MG
Sulfo-Cy3 Azide is a water-soluble fluorogenic dye featuring an azide group, which facilitates Click Chemistry. It is suitable for labeling delicate molecules, including proteins. The azide group allows it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules, as well as ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • Water-soluble fluorogenic dye: allows for use in aqueous environments.
  • Azide group: enables Click Chemistry reactions.
  • Versatile labeling: can be used to label sensitive molecules such as proteins.
  • Compatible with CuAAc: reacts with alkyne groups in copper-catalyzed azide-alkyne cycloaddition.
  • Compatible with SPAAC: undergoes ring strain-promoted alkyne-azide cycloaddition with DBCO or BCN groups.