Learn More
Medchemexpress LLC Sulfo-Cy3 Azide | 1801695-56-6 | C35H46N6O10S3 | 5 MG
SDP
Catalog No. 5000373434
Encompass_Preferred
Sulfo-Cy3 Azide is a water-soluble fluorogenic dye that contains an azide group, facilitating Click Chemistry. This dye is suitable for labeling sensitive molecules, such as proteins.
  • Can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules.
  • Can undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
Catalog No. 50-003-73434 Supplier Medchemexpress LLC Supplier No. HY1517105MG
Sulfo-Cy3 Azide is a water-soluble fluorogenic dye that contains an azide group, facilitating Click Chemistry. This dye is suitable for labeling sensitive molecules, such as proteins.
  • Can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules.
  • Can undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.