Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sulfo-Cy3 azide | 1801695-56-6 | MFCD30527437 | 99.6% | 806.97 | C35H46N6O10S3 | 10 MG
SDP

Catalog No. 5000286899
Encompass_Preferred

Sulfo-Cy3 Azide is a water-soluble Cy3 fluorescent dye bearing an azide functional group for bioorthogonal click chemistry. It is designed for labeling sensitive biomolecules, such as proteins, via copper-catalyzed azide-alkyne cycloaddition (CuAAC) or strain-promoted azide-alkyne cycloaddition (SPAAC). The dye exhibits excitation/emission around 550/570 nm and is supplied in research pack sizes.

  • Water-soluble cyanine dye suitable for aqueous labeling reactions.
  • Azide functional group enables CuAAC and SPAAC click chemistry.
  • Excitation/emission approximately 550/570 nm for bright fluorescence.
  • Soluble in DMSO at 50 mg/mL (requires sonication; use fresh DMSO).
  • High purity suitable for research applications.

Catalog No. 50-002-86899 Supplier Medchemexpress LLC Supplier No. HY15171010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfo-Cy3 Azide is a water-soluble Cy3 fluorescent dye bearing an azide functional group for bioorthogonal click chemistry. It is designed for labeling sensitive biomolecules, such as proteins, via copper-catalyzed azide-alkyne cycloaddition (CuAAC) or strain-promoted azide-alkyne cycloaddition (SPAAC). The dye exhibits excitation/emission around 550/570 nm and is supplied in research pack sizes.

  • Water-soluble cyanine dye suitable for aqueous labeling reactions.
  • Azide functional group enables CuAAC and SPAAC click chemistry.
  • Excitation/emission approximately 550/570 nm for bright fluorescence.
  • Soluble in DMSO at 50 mg/mL (requires sonication; use fresh DMSO).
  • High purity suitable for research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.