Learn More
Medchemexpress LLC Sulfo-Cy3 maleimide potassium | 1656990-68-9 | 93.35% | 776.96 | 1 MG
SDP
Catalog No. 5000350726
Encompass_Preferred
Sulfo-Cy3 maleimide potassium is a fluorescent dye featuring a maleimide functionality at the end of a short alkyl linker. It can be utilized for labeling antibodies and other labile proteins.
  • Fluorescent dye
  • Maleimide functionality
  • Used for labeling antibodies
  • Used for labeling labile proteins
Catalog No. 50-003-50726 Supplier Medchemexpress LLC Supplier No. HY1370981MG
Sulfo-Cy3 maleimide potassium is a fluorescent dye featuring a maleimide functionality at the end of a short alkyl linker. It can be utilized for labeling antibodies and other labile proteins.
  • Fluorescent dye
  • Maleimide functionality
  • Used for labeling antibodies
  • Used for labeling labile proteins