Learn More
Medchemexpress LLC Sulfo-Cy3-NHS disodium | 865.90 g/mol | C36H41N3Na2O13S3 | 10 MG
SDP
Catalog No. 5000297234
Encompass_Preferred
Sulfo-Cy3-NHS disodium is a water-soluble Cyanine3 (Cy3) derivative bearing an NHS ester for amine-reactive conjugation. The sulfonate groups increase aqueous solubility, making this reagent suitable for labeling proteins, peptides, and antibodies for fluorescence detection and imaging.
  • Water-soluble NHS ester for efficient labeling of primary amines.
  • Improved aqueous solubility due to sulfonate groups.
  • Suitable for conjugation to proteins, antibodies, and peptides.
  • Enables fluorescence detection and imaging applications.
  • Stable solid form for routine laboratory handling and storage per manufacturer instructions.
Catalog No. 50-002-97234 Supplier Medchemexpress LLC Supplier No. HYD1848A10MG
Sulfo-Cy3-NHS disodium is a water-soluble Cyanine3 (Cy3) derivative bearing an NHS ester for amine-reactive conjugation. The sulfonate groups increase aqueous solubility, making this reagent suitable for labeling proteins, peptides, and antibodies for fluorescence detection and imaging.
  • Water-soluble NHS ester for efficient labeling of primary amines.
  • Improved aqueous solubility due to sulfonate groups.
  • Suitable for conjugation to proteins, antibodies, and peptides.
  • Enables fluorescence detection and imaging applications.
  • Stable solid form for routine laboratory handling and storage per manufacturer instructions.