Learn More
Medchemexpress LLC Sulfo-Cy5.5 Azide | 1955527-41-9 | C44H50N6O13S4 | 5 MG
SDP
Catalog No. 5000373435
Encompass_Preferred
Sulfo-Cy5.5 Azide is a click chemistry reagent that contains an azide group. It is a water-soluble dye with excitation at 673 nm and emission at 707 nm. It is designed to label sensitive molecules like peptides, proteins, and oligonucleotides. The azide group allows it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups, and ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • Click chemistry reagent
  • Contains an azide group
  • Water-soluble dye (Ex=673 nm, Em=707 nm)
  • Designed to label sensitive molecules such as peptides, proteins, and oligonucleotides
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules
  • Can undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN-containing molecules
Catalog No. 50-003-73435 Supplier Medchemexpress LLC Supplier No. HY1517115MG
Sulfo-Cy5.5 Azide is a click chemistry reagent that contains an azide group. It is a water-soluble dye with excitation at 673 nm and emission at 707 nm. It is designed to label sensitive molecules like peptides, proteins, and oligonucleotides. The azide group allows it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups, and ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
  • Click chemistry reagent
  • Contains an azide group
  • Water-soluble dye (Ex=673 nm, Em=707 nm)
  • Designed to label sensitive molecules such as peptides, proteins, and oligonucleotides
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules
  • Can undergo ring strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN-containing molecules