Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sulfo-Cy5 azide | 1782950-80-4 | 99.0% | 833.01 | C37H48N6O10S3 | 5 MG
SDP

Catalog No. 5000221500
Encompass_Preferred

Sulfo-Cy5 azide is a water-soluble, azide-functionalized Cy5 fluorescent dye designed for bioconjugation and imaging applications. It is used for labeling biomolecules (including RNA) via click chemistry and provides bright red/far-red fluorescence suitable for microscopy and flow cytometry.

  • Water-soluble Cy5 fluorescent dye.
  • Contains an azide group for click chemistry (CuAAC and SPAAC).
  • Excitation 645 nm, emission 670 nm.
  • High purity suitable for research applications.
  • Solid form; store at 4°C and protect from light.
  • Supplied as a 5 mg amount for small-scale experiments.

Catalog No. 50-002-21500 Supplier Medchemexpress LLC Supplier No. HY1457465MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfo-Cy5 azide is a water-soluble, azide-functionalized Cy5 fluorescent dye designed for bioconjugation and imaging applications. It is used for labeling biomolecules (including RNA) via click chemistry and provides bright red/far-red fluorescence suitable for microscopy and flow cytometry.

  • Water-soluble Cy5 fluorescent dye.
  • Contains an azide group for click chemistry (CuAAC and SPAAC).
  • Excitation 645 nm, emission 670 nm.
  • High purity suitable for research applications.
  • Solid form; store at 4°C and protect from light.
  • Supplied as a 5 mg amount for small-scale experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.