Learn More
Lumiprobe Sulfo-Cyanine5 azide, 1mg
Catalog No. NC0602812
Encompass_Preferred
Molecular formula: C35H43N6KO7S2, Molecular weight: 762.98, Appearance: dark blue crystals, Solubility: very high in water; Water-soluble red emitting fluorescent dye (an analog of Cy5) with with an azido group for click chemistry reactions.
Catalog No. NC0602812 Supplier Lumiprobe Supplier No. A3330
Molecular formula: C35H43N6KO7S2, Molecular weight: 762.98, Appearance: dark blue crystals, Solubility: very high in water; Water-soluble red emitting fluorescent dye (an analog of Cy5) with with an azido group for click chemistry reactions.