Learn More
Medchemexpress LLC Sulfo-Cy5-N3 | 1481447-40-8 | 99.1% | 724.89 g/mol | C35H44N6O7S2 | 5 MG
SDP
Catalog No. 5000281054
Encompass_Preferred
Sulfo-Cy5-N3 is a sulfonated Cy5 fluorescent dye bearing an azide functional group for bioorthogonal click labeling. The dye is water-soluble and can be conjugated to alkyne-containing molecules via copper-catalyzed azide-alkyne cycloaddition (CuAAC) or by strain-promoted azide-alkyne cycloaddition (SPAAC), producing strong red fluorescence suitable for biomolecule imaging (Ex 642 nm / Em 670 nm).
  • Water-soluble azide-functional fluorescent dye for bioorthogonal labeling.
  • Compatible with CuAAC and SPAAC click chemistries for versatile conjugation.
  • Red fluorescence with excitation/emission at 642/670 nm for imaging.
  • High purity suitable for sensitive labeling applications.
  • Store protected from light at low temperature for stability.
Catalog No. 50-002-81054 Supplier Medchemexpress LLC Supplier No. HYW4009255MG
Sulfo-Cy5-N3 is a sulfonated Cy5 fluorescent dye bearing an azide functional group for bioorthogonal click labeling. The dye is water-soluble and can be conjugated to alkyne-containing molecules via copper-catalyzed azide-alkyne cycloaddition (CuAAC) or by strain-promoted azide-alkyne cycloaddition (SPAAC), producing strong red fluorescence suitable for biomolecule imaging (Ex 642 nm / Em 670 nm).
  • Water-soluble azide-functional fluorescent dye for bioorthogonal labeling.
  • Compatible with CuAAC and SPAAC click chemistries for versatile conjugation.
  • Red fluorescence with excitation/emission at 642/670 nm for imaging.
  • High purity suitable for sensitive labeling applications.
  • Store protected from light at low temperature for stability.