Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sulfo-Cy5-N3 | 1481447-40-8 | MFCD28968783 | 99.1% | 724.89 | C35H44N6O7S2 | 25 MG
SDP

Catalog No. 5000281008
Encompass_Preferred

Sulfo-Cy5-N3 is a sulfonated Cyanine5 fluorescent azide reagent for click chemistry labeling of biomolecules. It delivers bright red fluorescence for imaging, improved aqueous solubility compared with non-sulfonated Cy5, and is supplied as a research-grade solid. Handle protected from light and prepare stock solutions in dry DMSO according to recommended storage conditions.

  • Provides bright red fluorescence with Ex/Em 642/670 nm.
  • Contains an azide group for click chemistry labeling of biomolecules.
  • Sulfonation improves aqueous solubility compared with non-sulfonated Cy5.
  • Soluble in DMSO up to 125 mg/mL; ultrasonic bath may aid dissolution.
  • High purity suitable for research use (99.1%).
  • Store protected from light at -20°C; in solution store at -80°C up to 6 months.

Catalog No. 50-002-81008 Supplier Medchemexpress LLC Supplier No. HYW40092525MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfo-Cy5-N3 is a sulfonated Cyanine5 fluorescent azide reagent for click chemistry labeling of biomolecules. It delivers bright red fluorescence for imaging, improved aqueous solubility compared with non-sulfonated Cy5, and is supplied as a research-grade solid. Handle protected from light and prepare stock solutions in dry DMSO according to recommended storage conditions.

  • Provides bright red fluorescence with Ex/Em 642/670 nm.
  • Contains an azide group for click chemistry labeling of biomolecules.
  • Sulfonation improves aqueous solubility compared with non-sulfonated Cy5.
  • Soluble in DMSO up to 125 mg/mL; ultrasonic bath may aid dissolution.
  • High purity suitable for research use (99.1%).
  • Store protected from light at -20°C; in solution store at -80°C up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.