Learn More
Apexbio Technology LLC SULFO-CY5-N3-5MG
SDP
Catalog No. 5000569379
Encompass_Preferred
Sulfo-Cy5-N3 (CAS No 1481447-40-8) is a water-soluble fluorescent reagent commonly utilized in bioimaging and labeling applications within the field of biomedical research Originating from the family of cyanine dyes it targets biomolecular structures through azide-alkyne cycloaddition also known as click chemistry allowing for specific labeling of alkyne-modified molecules Its in vitro activity is demonstrated in the nanomolar to low micromolar range making it highly effective for precise applications Sulfo-Cy5-N3 is widely used in cell models animal studies and drug screening due to its stable fluorescence and compatibility with various biological systems Experimental concentrations typically depend on the specific research design enabling researchers to adapt usage according to the requirements of their studies
Catalog No. 50-005-69379 Supplier Apexbio Technology LLC Supplier No. C82925
Sulfo-Cy5-N3 (CAS No 1481447-40-8) is a water-soluble fluorescent reagent commonly utilized in bioimaging and labeling applications within the field of biomedical research Originating from the family of cyanine dyes it targets biomolecular structures through azide-alkyne cycloaddition also known as click chemistry allowing for specific labeling of alkyne-modified molecules Its in vitro activity is demonstrated in the nanomolar to low micromolar range making it highly effective for precise applications Sulfo-Cy5-N3 is widely used in cell models animal studies and drug screening due to its stable fluorescence and compatibility with various biological systems Experimental concentrations typically depend on the specific research design enabling researchers to adapt usage according to the requirements of their studies