Learn More
Lumiprobe Sulfo-Cyanine5 NHS ester, 2230212-27-6, 5mg
Catalog No. NC0216682
Encompass_Preferred
Molecular formula: C36H40N3KO10S2, Molecular weight: 777.95, Appearance: dark blue powder, Solubility: very good in water, good in DMF and DMSO; Water-soluble red emitting fluorescent dye. Amine-reactive Cy5 analog for the labeling of proteins/peptides and amino-oligonucleotides.
Catalog No. NC0216682 Supplier Lumiprobe Supplier No. 23320
Molecular formula: C36H40N3KO10S2, Molecular weight: 777.95, Appearance: dark blue powder, Solubility: very good in water, good in DMF and DMSO; Water-soluble red emitting fluorescent dye. Amine-reactive Cy5 analog for the labeling of proteins/peptides and amino-oligonucleotides.