Learn More
Medchemexpress LLC Sulfo-Cy7.5 NHS ester | 2736437-44-6 | 97.0% | 1104.29 g/mol | C49H48K3N3O16S4 | 10 MG
SDP
Catalog No. 5000297819
Encompass_Preferred
Sulfo-Cy7.5 NHS ester is a water-soluble, hydrophilic near-infrared cyanine dye functionalized as an N-hydroxysuccinimide (NHS) ester for covalent labeling of primary amines on proteins, peptides, and other biomolecules.
  • Near-infrared, water-soluble cyanine dye suitable for NIR imaging and fluorescence labeling.
  • Reactive NHS ester for efficient covalent coupling to primary amines under mild conditions.
  • Reported molecular weight of 1104.29 g/mol for planning conjugation ratios.
  • High reported purity suitable for research applications and labeling workflows.
  • Available in small milligram pack sizes for pilot studies and labeling optimization.
  • Store sealed, away from moisture and light to maintain stability.
Catalog No. 50-002-97819 Supplier Medchemexpress LLC Supplier No. HYD1568A10MG
Sulfo-Cy7.5 NHS ester is a water-soluble, hydrophilic near-infrared cyanine dye functionalized as an N-hydroxysuccinimide (NHS) ester for covalent labeling of primary amines on proteins, peptides, and other biomolecules.
  • Near-infrared, water-soluble cyanine dye suitable for NIR imaging and fluorescence labeling.
  • Reactive NHS ester for efficient covalent coupling to primary amines under mild conditions.
  • Reported molecular weight of 1104.29 g/mol for planning conjugation ratios.
  • High reported purity suitable for research applications and labeling workflows.
  • Available in small milligram pack sizes for pilot studies and labeling optimization.
  • Store sealed, away from moisture and light to maintain stability.