Learn More
Medchemexpress LLC Sulfo-Cy7-DBCO | 95.1% | 10 MG
SDP
Catalog No. 5000297564
Encompass_Preferred
Sulfo-Cy7-DBCO is a water-soluble derivative of cyanine 7 bearing a DBCO moiety for strain-promoted alkyne-azide cycloaddition (SPAAC). It provides near-infrared fluorescence with spectral properties comparable to Cy7 and is intended for bioconjugation and imaging applications.
  • Water-soluble near-infrared fluorescent dye.
  • DBCO functionality enables copper-free click conjugation (SPAAC).
  • Spectral properties similar to Cy7 for NIR imaging compatibility.
  • Provided at high purity (~95.1%).
  • Supplied in small-mass quantities suitable for labeling workflows.
  • Store sealed, away from moisture and light at 4°C; in solvent store at -80°C or -20°C as recommended.
Catalog No. 50-002-97564 Supplier Medchemexpress LLC Supplier No. HYD224110MG
Sulfo-Cy7-DBCO is a water-soluble derivative of cyanine 7 bearing a DBCO moiety for strain-promoted alkyne-azide cycloaddition (SPAAC). It provides near-infrared fluorescence with spectral properties comparable to Cy7 and is intended for bioconjugation and imaging applications.
  • Water-soluble near-infrared fluorescent dye.
  • DBCO functionality enables copper-free click conjugation (SPAAC).
  • Spectral properties similar to Cy7 for NIR imaging compatibility.
  • Provided at high purity (~95.1%).
  • Supplied in small-mass quantities suitable for labeling workflows.
  • Store sealed, away from moisture and light at 4°C; in solvent store at -80°C or -20°C as recommended.