Learn More
Lumiprobe Sulfo-Cyanine7 NHS ester, 1603861-95-5, 5mg
Catalog No. NC0494171
Encompass_Preferred
Molecular formula: C41H46N3KO10S2, Molecular weight: 844.05, Appearance: dark green powder, Solubility: good in water, DMF, DMSO; Sulfo-Cyanine7 is a water-soluble near infrared dye. The fluorophore is ideal for the labeling of complex proteins for subsequent live organism NIR imaging studies or sonoluminescence.
Catalog No. NC0494171 Supplier Lumiprobe Supplier No. 25320
Molecular formula: C41H46N3KO10S2, Molecular weight: 844.05, Appearance: dark green powder, Solubility: good in water, DMF, DMSO; Sulfo-Cyanine7 is a water-soluble near infrared dye. The fluorophore is ideal for the labeling of complex proteins for subsequent live organism NIR imaging studies or sonoluminescence.